Sodium-L(+)-glutamate monohydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6106-04-3
:
Formula:C₅H₈NNaO₄·H₂O
MDL Number:MFCD00150138
:187.13 g/mol
Merck Index:13,06278
Melting Pt:228 °C (dec.)
Flash Pt:>225 °C
Storage Temperature:Ambient
EINECS:205-538-1
Related Information
Frequently Bought Together