Order Entry
Italy
ContactUsLinkComponent
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid 98%
Irritant
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid 98%
Catalog # APOSPC501601-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid 98%
Catalog # APOSPC501601-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.92 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:190903-71-0
Formula:C₇H₅BF₂O₄
MDL Number:MFCD08706236