3,5-Difluoro-4-(trifluoromethyl)benzaldehyde
:1417569-98-2
:
:C₈H₃F₅O
:MFCD22566356
:210.1 g/mol
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals