
4-Bromo-3-(difluoromethyl)benzyl bromide 95%
Catalog # APOSPC57769-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:299.94 g/mol
UN:3265
ADR:8,II
:GHS05
CAS Number:1261580-21-5
Formula:C₈H₆Br₂F₂
MDL Number:MFCD18391075