
2-Bromo-5-(difluoromethyl)benzonitrile 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1261441-87-5
:
Formula:C₈H₄BrF₂N
MDL Number:MFCD18391059
:232.02 g/mol
UN:3439
ADR:6.1,III
:GHS06
Frequently Bought Together
Cyclohexane, EMPLURA®, Supelco®
Supplier: Merck
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
1-Propanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Dichloromethane ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dibutylamine for HPLC
Supplier: Thermo Fisher Scientific