Order Entry
Italy
ContactUsLinkComponent
Methyl 2,3-difluoro-6-(trifluoromethoxy)benzoate 95%
Irritant
Methyl 2,3-difluoro-6-(trifluoromethoxy)benzoate 95%
Catalog # APOSPC57464-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 2,3-difluoro-6-(trifluoromethoxy)benzoate 95%
Catalog # APOSPC57464-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.13 g/mol
 :GHS07
CAS Number:2703797-04-8
Formula:C₉H₅F₅O₃