Order Entry
Italy
ContactUsLinkComponent
4-Bromo-2-fluoro-3-(trifluoromethyl)phenacyl bromide 95%
Corrosive
4-Bromo-2-fluoro-3-(trifluoromethyl)phenacyl bromide 95%
Catalog # APOSPC57424-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
4-Bromo-2-fluoro-3-(trifluoromethyl)phenacyl bromide 95%
Catalog # APOSPC57424-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :363.93 g/mol
UN:3261
ADR:8,II
 :GHS05
CAS Number:2386358-17-2
Formula:C₉H₄Br₂F₄O
MDL Number:MFCD34181535