
2-Bromo-4-(trifluoromethoxy)benzyl alcohol 97%
:1316273-53-6
:
:C₈H₆BrF₃O₂
:MFCD19105411
:271.031 g/mol
:GHS07
Frequently Bought Together
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Isobutyronitrile 99%
: Thermo Fisher Scientific
Glycerol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 2,5 L
: PanReac AppliChem
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
: Thermo Fisher Scientific
Methyl methanesulphonate 99%
: Thermo Fisher Scientific
Toluene ≥99.5% ACS
: Thermo Fisher Scientific
Methanol bio-based ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
: PanReac AppliChem