
3'-(1,1-Difluoroethyl)-2,2,2-trifluoroacetophenone 95%
:
:C₁₀H₇F₅O
:238.15 g/mol
:GHS07
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
: MERCK PRODUCTION CHEMICALS
Water, extra pure, Deionized
: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.5%, Reagent Grade
: Honeywell Chemicals
Water, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
1-Pentanol 99+% ACS
: Thermo Fisher Scientific
1-Pentanol 99%, pure
: Thermo Fisher Scientific
1,4-Dioxane 99+% ACS
: Thermo Fisher Scientific
Methyl formate 98% for spectroscopy
: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
: PanReac AppliChem
Chloroform 99+% stabilised, extra pure
: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99%
: Thermo Fisher Scientific