
N-(1-Methyl-1H-indazol-3-yl)thiourea
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2594436-05-0
:
Formula:C₉H₁₀N₄S
MDL Number:MFCD32857216
:206.27 g/mol
:GHS07
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.5% (by GC), EMPROVE® ESSENTIAL NF, Ph. Eur., BP, JPE, extra pure suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Pyridine ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethyl methanesulphonate 99%
Supplier: Thermo Fisher Scientific
Sinapic acid, Supelco®
Supplier: Merck
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific