Order Entry
Italy
ContactUsLinkComponent
2,3-Difluoro-6-(trifluoromethoxy)benzaldehyde 95%
2,3-Difluoro-6-(trifluoromethoxy)benzaldehyde 95%
Catalog # APOSPC502497-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Difluoro-6-(trifluoromethoxy)benzaldehyde 95%
Catalog # APOSPC502497-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :226.1 g/mol
CAS Number:2244081-97-6
Formula:C₈H₃F₅O₂
MDL Number:MFCD31561166