
2,2'-Dibromobiphenyl (PBB No. 4) 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13029-09-9
:
Formula:C₁₂H₈Br₂
MDL Number:MFCD00093707
:312 g/mol
Boiling Pt:252…274 °C
Melting Pt:78…83 °C
Density:1.81
:GHS07
Frequently Bought Together