Order Entry
Italy
ContactUsLinkComponent
(±)-1-Aminopropan-2-ol 95%
(±)-1-Aminopropan-2-ol 95%
Catalog # APOSOR923138-100G
Supplier:  Apollo Scientific
CAS Number:  
(±)-1-Aminopropan-2-ol 95%
Catalog # APOSOR923138-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :75.11 g/mol
Flash Pt:105 °C
UN:2735
ADR:8,III
 :GHS05
CAS Number:78-96-6
Formula:C₃H₉NO
MDL Number:MFCD00008139