
4-Hydroxy-3-(trifluoromethyl)benzaldehyde
Catalog # APOSPC3247Q-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:190.12 g/mol
Melting Pt:158 - 165 °C
:GHS07
CAS Number:220227-98-5
Formula:C₈H₅F₃O₂
MDL Number:MFCD01091006
Melting Pt:158…165 °C