
4-Bromo-2-fluoro-5-(trifluoromethyl)benzaldehyde 95%
:1414870-67-9
:
:C₈H₃BrF₄O
:MFCD22628190
:271.006 g/mol
:GHS07
Frequently Bought Together
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
: VWR Chemicals
Carbon disulphide 99.9% for HPLC
: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Butyl acetate ≥99.5%, Puriss. p.a. ACS, Riedel-de Haën™
: Honeywell Chemicals
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
1,4-Dioxane ≥99% for HPLC
: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
Methanol (USP-NF, BP, Ph. Eur.) pure, pharma grade 1 * 2,5 L
: PanReac AppliChem
2-Butoxyethanol ≥99%
: Thermo Fisher Scientific
Cyclohexane 99.8% for HPLC
: Thermo Fisher Scientific
Ethylbenzene 99.8%, pure
: Thermo Fisher Scientific
Diethyl ether 99.5% stabilised, meets analytical specification of Ph.Eur. for analysis
: Thermo Fisher Scientific