Order Entry
Italy
ContactUsLinkComponent
Methyl 4-bromo-2-iodo-5-methoxybenzoate 95%
Methyl 4-bromo-2-iodo-5-methoxybenzoate 95%
Catalog # APOSOR62216-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-bromo-2-iodo-5-methoxybenzoate 95%
Catalog # APOSOR62216-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :370.97 g/mol
CAS Number:2384857-60-5
Formula:C₉H₈BrIO₃