Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, Cumene 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, Cumene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA14463500ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Cumene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA14463500ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    98-82-8
  • Description:
    Cumene 100 µg/ml in methanol
  • Formula:
    C₉H₁₂
  • Description:
    Cumene

Specifications

About this item