Pesticide reference standard, 2,3,5,6-Tetrachloroanisole 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL17333300IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:6936-40-9
- Description:2,3,5,6-Tetrachloroanisole 10 µg/ml in isooctane
- Formula:C₇H₄Cl₄O
- Description:2,3,5,6-Tetrachloroanisole