Organic reference standard, cis-Dichloroethylene 200 µg/ml in methanol, Supelco®
Catalog # 48836.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- CAS No.:156-59-2
- Description:cis-Dichloroethylene 200 µg/ml in methanol
- Formula:C₂H₂Cl₂
- Description:cis-Dichloroethylene