Lecithin from egg yolk, GPR RECTAPUR®
Catalog # 290534N
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- N (Nitrogen):1.4 - 1.9 %
- P (Phosphorus):About 3 %
- Loss on drying:Max. 2 %
Specification Test Results
About this item
:
EINECS:297-639-2
CAS Number:93685-90-6
MDL Number:MFCD00131449