Gibberellic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:77-06-5
:
Formula:C₁₉H₂₂O₆
MDL Number:MFCD00079329
:346.38 g/mol
Merck Index:13,04430
Melting Pt:234 °C
Density:1.386 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:201-001-0
Specification Test Results
- Solubility:5 g/l (25 °C)
- Melting Point:233 - 235 °C
- Molar Mass:346.37 g/mol
- Density:1.34 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together