Dansyl chloride ≥95%
Plastic bottle, 100 mg
Catalog # AATB811
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Dansyl chloride is non-fluorescent until it reacts with amines.
Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride
- CAS Number:605-65-2
- Formula:C₁₂H₁₂ClNO₂S
- MDL Number:MFCD00003985
- Molecular Weight:269.75 g/mol
- ADR:5,II
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:100 mg



