Order Entry
Italy
ContactUsLinkComponent
 
 
DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)] ≥95%
  AATB62
 :  
undefined
 
DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)] ≥95%
  AATB62
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

 

 

 :
 :308.16 g/mol
 :5,II
 :GHS07
 :118290-05-4
 :C₁₅H₁₁Cl₂NO₂
 :MFCD01311087