Order Entry
Italy
ContactUsLinkComponent
6-Carboxy-2',4,7',7-tetrachlorofluorescein succinimidyl ester (6-TET SE) ≥90%
6-Carboxy-2',4,7',7-tetrachlorofluorescein succinimidyl ester (6-TET SE) ≥90%
Catalog # AATB211
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
6-Carboxy-2',4,7',7-tetrachlorofluorescein succinimidyl ester (6-TET SE) ≥90%
Catalog # AATB211
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :611.17 g/mol
Formula:C₂₅H₁₁Cl₄NO₉