
Fluorescein-12-dUTP sodium salt 1 mM in TRIS buffer ≥95% pH 7.5
Codice catalogo AATB17022
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 Nmo
- Seq:0311
Specifications
About this item
:
:1060.62 g/mol
ADR:5,II
:GHS07
CAS Number:214154-36-6
Formula:C₃₉H₃₈N₄O₂₁P₃Na₃