Order Entry
Italy
Orders LinkContactUsLinkComponent
DAX-J2™ Red

DAX-J2™ Red

Plastic bottle, 1 mg
Catalog # AATB16301
Irritant
Supplier:   AAT Bioquest
DAX-J2™ Red
Catalog # AATB16301
Supplier:  AAT Bioquest
CAS Number:  
DAX-J2™ Red
DAX-J2™ Red

Plastic bottle, 1 mg

CAS Number:  
Price
235,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

DAX-J2™ Red is a new nitric oxide (NO) sensor recently developed by AAT Bioquest.

  • Does not require esterase activity for no detection
  • Ph-independent fluorescence
  • More photostable than daf-2 product
  • More sensitive for detecting no than daf-2

Caution: For research use only.

  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications