Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
UV375 succinimidyl ester
UV375 succinimidyl ester, mFluor™ fluorescent probe
UV375 succinimidyl ester, mFluor™ fluorescent probe
AATB1135
:
AAT BIOQUEST
:
UV375 succinimidyl ester, mFluor™ fluorescent probe
AATB1135
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
782.95 g/mol
:
5,II
:
GHS07