LysoBrite™ Red
LysoBrite™ red, 500 Tests
Catalog # AATB22645
Supplier: AAT Bioquest
Jump to:
Product Details & Documents
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
- Lysosome-specific dye
- Long-term use
- Low cytotoxicity
Caution: For research use only.
Specifications
Specifications
- Pk:500 Tests
- Description:LysoBrite™ red
- Cat. No.:AATB22645
- Purity:95%