Cell Navigator® Mitochondrion Staining Kit *Green Fluorescence*
Catalog # AATB22666
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:500 Assays
- Description:Cell Navigator™ mitochondrion staining kit *green fluorescence*
Specifications
About this item
Our Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labelling sub-cellular organelles such as membranes, lysosomes, mitochondria and nuclei etc.
- Cell-permeant staining
- Improved signal
- Suitable for all major fluorescence platforms
Caution: For research use only.