Buccutite™ Peroxidase (HRP) Antibody Conjugation Kits (2 Labelings)
Supplier: AAT BIOQUEST
Certificates
About this item
Protein-protein conjugations are commonly performed with a bifunctional linker (such as the commonly used SMCC), having different reactivity on each end for linking two different proteins.
- High conjugation yield
- Stable crosslinking
- Minimal hands-on time
- Robust protocol
Related Information
Manual Buccutite peroxidase Hrp Antibody Conjµgation Kit Optimized for Labeling 25 µg Protein.pdf Manual Buccutite™ Peroxidase Hrp Antibody Conjµgation Kit Optimized For Labeling 1 Mg Protein.pdf Manual Buccutite™ Peroxidase Hrp Antibody Conjµgation Kit Optimized For Labeling 100 µg Protein.pdf Manual Buccutite™Peroxidase (HRP) Antibody Conjugation Kit Optimized for Labeling 1 mg Protein.pdf Manual Peroxidase (HRP) Antibody Conjugation Kit (2 Labelings).pdf
Manual
More
Frequently Bought Together