Order Entry
Italy
ContactUsLinkComponent
Cell Meter™ Flow Cytometric Calcium Assay Kit
Cell Meter™ Flow Cytometric Calcium Assay Kit
Catalog # AATB36310
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ Flow Cytometric Calcium Assay Kit
Catalog # AATB36310
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ flow cytometric calcium assay kit

Specifications

About this item

Cell Meter™ flow cytometric calcium assay kit provides fluorescence-based assays for detecting intracellular calcium mobilisation using a flow cytometer.

  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Optimised for flow cytometers but suitable for all major fluorescence platforms
  • Multiple research applications
Caution: For research use only.