Amplite® Fluorimetric D-Lactate Assay Kit
Catalog # AATB13810
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:Amplite™ fluorimetric D-lactate assay kit
Specifications
About this item
Lactic acid is chiral and has two optical isomers: L-lactic acid and D-lactic acid.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.