Order Entry
Italy
ContactUsLinkComponent
Cell Meter™ Caspase 9 Activity Apoptosis Assay Kit *Blue Fluorescence*
Cell Meter™ Caspase 9 Activity Apoptosis Assay Kit *Blue Fluorescence*
Catalog # AATB22813
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ Caspase 9 Activity Apoptosis Assay Kit *Blue Fluorescence*
Catalog # AATB22813
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Cell Meter™ caspase 9 activity apoptosis assay kit *blue fluorescence*

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cellular functions.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.