Amplite® Fluorimetric Acetylcholinesterase Assay Kit *Green Fluorescence*
Catalog # AATB11401
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:200 Tests
- Description:Amplite™ fluorimetric acetylcholinesterase assay kit *green fluorescence*
- Tests per kit:200 Tests
Specifications
About this item
Acetylcholinesterase, also known as AChE, is an enzyme that degrades (through its hydrolytic activity) the neurotransmitter acetylcholine, producing choline and an acetate group.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.