MESG *Phosphate Assay Reagent*
Catalog # AATB21600
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:5 mg
- Description:MESG *Phosphate assay reagent*
Specifications
About this item
In the presence of inorganic phosphate MESG is converted to 2-amino-6-mercapto-7-methlpurine by purine nucleoside phosphorylase (EC 2.4.2.1) with absorption wavelength shift to red.
- Adaptable and robust
- Quantifiable emission wavelength shift
- Standalone reagent
Caution: For research use only.