Cell Meter™ Cell Viability Assay Kit *Blue Fluorescence with 405 nm Excitation*
Catalog # AATB22784
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:500 Tests
- Description:Cell Meter™ cell viability assay kit *blue fluorescence with 405 nm excitation*
- Tests per kit:500 Tests
Specifications
About this item
There are a variety of parameters that can be used to monitor cell viability.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Improved detection sensitivity
Caution: For research use only.