Order Entry
Italy
ContactUsLinkComponent
Cell Navigator® Lysosome Staining Kit *Red Fluorescence*
Cell Navigator® Lysosome Staining Kit *Red Fluorescence*
  AATB22658
 :  
undefined
Cell Navigator® Lysosome Staining Kit *Red Fluorescence*
  AATB22658
 :  
  • Pk:
    500 Tests
  • Description:
    Cell Navigator™ lysosome staining kit *red fluorescence*
  • Tests per kit:
    500 Tests
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
  • Cell-permeant staining
  • Improved signal
  • Suitable for all major fluorescence platforms
 : For research use only.