Cell Explorer™ Live Cell Labelling Kit *Blue Fluorescence*
Catalog # AATB22606
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:Cell Explorer™ live cell labeling kit *blue fluorescence*
- Tests per kit:200 Tests
Specifications
About this item
Our Cell Explorer™ fluorescence imaging kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
- Results in under 2 hours
Caution: For research use only.