Cell Meter™ Intracellular Colorimetric Lipid Peroxidation (MDA) Assay Kit
AATB15991
:
- Pk:200 Tests
- Description:Cell Meter™ Intracellular colorimetric lipid peroxidation (MDA) assay kit
- Tests per kit:200 Tests
Lipid peroxidation is characterised by the oxidative degradation of unsaturated fatty acids, phospholipids, glycolipids, cholesterol esters and cholesterol.
- MDA-specific
- Robust protocol
- No high temperatures required
: For research use only.