Order Entry
Italy
ContactUsLinkComponent
MycoLight™ Green JJ98 *5 mM in DMSO*
MycoLight™ Green JJ98 *5 mM in DMSO*
Catalog # AATB24000
Supplier:  AAT Bioquest
CAS Number:  
undefined
MycoLight™ Green JJ98 *5 mM in DMSO*
Catalog # AATB24000
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Description:
    MycoLight™ Green JJ98
  • Purity:
    95%

Specifications

About this item

MycoLight™ Green JJ98 is a green-fluorescent nuclear and chromosome stain that is permeant to both prokaryotic and eukaryotic cell membranes.

  • Cell-Permeable
  • High DNA Affinity
  • Bright Fluorescence
Caution: For research use only.