MitoLite™ Green FM
Catalog # AATB22695
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:500 µG
- Description:MitoLite™ Green FM
- Purity:90%
Specifications
About this item
MitoLite™ Green FM is the same molecule to the MitoTracker Green FM (M7514, ThermoFisher).
- Mitochondria-Specific Dye
- Cell-Permeable
- Robust
Caution: For research use only.