Order Entry
Italy
Orders LinkContactUsLinkComponent
Cell Meter™ Fixable Viability Dye, VX500
Cell Meter™ Fixable Viability Dye, VX500
Cell Meter™ VX500 fixable cell stain, 200 Tests, 200 Tests
Codice catalogo AATB22542
Nome del fornitore:   AAT Bioquest
Cell Meter™ Fixable Viability Dye, VX500
Codice catalogo AATB22542
Nome del fornitore:  AAT Bioquest
Numero CAS:  
Cell Meter™ Fixable Viability Dye, VX500
Cell Meter™ Fixable Viability Dye, VX500
Cell Meter™ VX500 fixable cell stain, 200 Tests, 200 Tests
Numero CAS:  
Price
299,00 €
Log in for availability
1 * 200 Tests
Product Details & Documents
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
  • Cell-Impermeable
  • Amine- And Thiol-Reactive
  • High Stability
  • Live/Dead Cell Labeling Dye
  • Fixable
Caution: For research use only.
Specifications
Specifications
  • Pk:
    200 Tests
  • Description:
    Cell Meter™ VX500 fixable cell stain
  • Tests per kit:
    200 Tests
Specifications