Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Nuclear Blue™ DCS1

Nuclear Blue™ DCS1, 0,5 mL
Catalog # AATB17548
Supplier:   AAT Bioquest
Nuclear Blue™ DCS1
Catalog # AATB17548
Supplier:  AAT Bioquest
CAS Number:  
Nuclear Blue™ DCS1
Nuclear Blue™ DCS1

Nuclear Blue™ DCS1, 0,5 mL

CAS Number:  
Price
156,00 €
Log in for availability
1 * 0,5 mL
Product Details & Documents

Our Nuclear Blue™ DCS1 is a fluorogenic, DNA-selective and cell-impermeant dye for analyzing DNA content in dead, fixed or apoptotic cells.

  • Cell-Impermeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence

5 mM DMSO Solution

 : For research use only.

Specifications

Specifications

  • Pk:
    0,5 mL
  • Description:
    Nuclear Blue™ DCS1
  • Cat. No.:
    AATB17548
  • Purity:
    95%

Specifications