1,3-Propanediol ≥99.7%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:504-63-2
:
Formula:HO(CH₂)₃OH
MDL Number:MFCD00002949
:76.1 g/mol
Merck Index:12,09845
Boiling Pt:214 °C (1013 hPa)
Melting Pt:–32 °C
Density:1.0597 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-997-3
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating)
Specifications
Specification Test Results
- Assay:Min. 99.7 %
- IR Spectrum:Passes test
- Colouration:Max. 15 APHA
- Water:Max. 0.05 %
Specification Test Results
Related Information
Frequently Bought Together