Order Entry
Italy
ContactUsLinkComponent
Nile blue A ≥95%
Irritant
Nile blue A ≥95%
Catalog # AATB60
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Nile blue A ≥95%
Catalog # AATB60
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :732.84 g/mol
ADR:5,II
 :GHS07
CAS Number:3625-57-8
Formula:C₄₀H₄₀N₆O₆S
MDL Number:MFCD00064529