Order Entry
Italy
ContactUsLinkComponent
6-Chloro-8-fluoro-umbelliferone fluorescence reference standard
6-Chloro-8-fluoro-umbelliferone fluorescence reference standard
Catalog # AATB42
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
6-Chloro-8-fluoro-umbelliferone fluorescence reference standard
Catalog # AATB42
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
ADR:5,II
 :228.6 g/mol
 :GHS07