Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Carboxy-4,7-dichloror-X-hodamine (6-dROX)
6-Carboxy-4,7-dichloror-X-hodamine (6-dROX)
6-Carboxy-4,7-dichloror-X-hodamine (6-dROX)
6-Carboxy-4,7-dichloror-X-hodamine (6-dROX)
Catalog # AATB311
Supplier:
AAT Bioquest
CAS Number:
Expand image
6-Carboxy-4,7-dichloror-X-hodamine (6-dROX)
Catalog # AATB311
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
:
:
704.69 g/mol
Product Details