Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Cal-520FF™ AM

Irritant
Supplier:   AAT Bioquest
Cal-520FF™ AM
Supplier:  AAT Bioquest
Certificates
Cal-520FF™ AM

2 Options Available Below

Product Details & Documents

Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.

  • Improved background/signal ratio
  • Suitable for all major fluorescence platforms
  • Cell permeant

 : For research use only.

  • Molecular Weight:1138.92 g/mol
  • ADR:5,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG - 548,00 €
    548,00 €
    1 * 500 µG
       548,00 €

  • Plastic bottle
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 785,00 €
    785,00 €
    1 * 1 mg
       785,00 €