
Fura-10™ AM
Supplier: AAT BIOQUEST
Certificates
About this item
:
:1194.14 g/mol
ADR:5,II
:GHS07
Frequently Bought Together
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, sterile, polypropylene, Corning®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Spatulas
Supplier: VWR Collection
Laboratory trays
Supplier: KARTELL
VWR®, Spreaders, T-shaped
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Aluminium Foil Seals for PCR and Storage (96- and 384-Well Plates)
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR® Electroporation Cuvettes
Supplier: VWR Collection
Multiple well cell culture plates, Costar®
Supplier: Corning
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection