
6-(4,6-Dichlorotriazinyl)aminofluorescein
Catalog # AATB205
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:495.28 g/mol
ADR:5,II
:GHS07
CAS Number:118357-32-7
Formula:C₂₃H₁₂Cl₂N₄O₅